Reusable Maxima ESD Cleanroom Coveralls, Uniform Technology
Supplier: Uniform Technology
Certificates
About this item
Cleanroom coveralls are manufactured from 99% polyester/1% carbon fabric.
- Military collar
- Raglan sleeves
- ESD knit wrists
- Coil zipper with top snaps
- Snap closures at ankle and back of legs
Suitable for ISO Class 4 or higher cleanrooms.
Specifications
- Catalog No:
- 76515-900
- 76515-898
- 76515-896
- 76515-894
- 76515-892
- 76515-890
- 76515-888
- 76515-886
- 76515-884
- Model:
- CC1245-68
- CC1245-68
- CC1245-68
- CC1245-68
- CC1245-68
- CC1245-68
- CC1245-68
- CC1245-68
- CC1245-68
- Garment application:
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Risk area:
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Fabric:
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon