Vikan® Drain Cleaning Brush, Remco
Catalog # 76530-046
Supplier: Remco Products
Specifications
- Description:Drain cleaning brush, black
- Cat. no.:76530-046
Specifications
About this item
The Drain Cleaning Brush is great for cleaning drains and gullies with limited access.
- Polyester bristles, polypropylene grip
- Euro (hygienic) thread type
Its flexible, rounded head gives it the ability to clean drains of any size, while still being able to conform to the shape of gullies. It can be used with any Vikan Euro threaded handle.
Overall dimensions: 10.83L×3.78W×2.36"H