Apostle MiniMax™ Isolation Kit, Beckman Coulter
Cell-Free DNA (cfDNA) Isolation from Plasma. Apostle MiniMax™ High Efficiency Cell-Free DNA Isolation Kit isolates cell-free DNA (cfDNA) from plasma, serum, and urine. This kit is powered by paramagnetic bead-based technology and isolates DNA suitable for NGS and PCR-based downstream applications.
- Automated available
- Sample type:Plasma
Frequently Bought Together