Order Entry
Canada
ContactUsLinkComponent
SP Wilmad-LabGlass, Variable Temperature NMR Spinner Turbine for use with Bruker® Spectrometers, SP Industries
SP Wilmad-LabGlass, Variable Temperature NMR Spinner Turbine for use with Bruker® Spectrometers, SP Industries
Catalog # 76460-012
Supplier:  SP Industries
undefined
SP Wilmad-LabGlass, Variable Temperature NMR Spinner Turbine for use with Bruker® Spectrometers, SP Industries
Catalog # 76460-012
Supplier:  SP Industries
Supplier Number:  B-PEEK-5

Specifications

  • Description:
    NMR spinner turbine, fits tube size: 5 mm
  • Material:
    PEEK with Viton™ O-rings
  • Cat. no.:
    76460-012

Specifications

About this item

Manufactured to precision standards that exceed the spectrometer manufacturers requirements.

  • Variable temperature operation of −150 to +200 °C
  • Weight compatible with room temperature spinners
  • Susceptibility matched with Bruker® originals

Can be mixed with other turbines during sample changer operation.