ContactUsLinkComponent
Martin Lewis with Lincomycin, for Neisseria gonorrhoeae, Hardy Diagnostics
Martin Lewis with Lincomycin, for Neisseria gonorrhoeae, Hardy Diagnostics
Catalog # 76523-366
Supplier:  Hardy Diagnostics
CAS Number:  
undefined
Martin Lewis with Lincomycin, for Neisseria gonorrhoeae, Hardy Diagnostics
Catalog # 76523-366
Supplier:  Hardy Diagnostics
Supplier Number:  E39
CAS Number:  
Specifications
  • Description:
    Prepared Martin Lewis with Lincomycin
  • Size:
    15×100 mm plate
  • Cat. no.:
    76523-366
Specifications
About this item
For the selective isolation of Neisseria gonorrhoeae.
  • Martin Lewis Agar with lincomycin, reduced vancomycin, colistin, amphotericin, and trimethoprim
Hardy Diagnostics Martin Lewis with Lincomycin is a selective medium for the recovery of Neisseria gonorrhoeae from both genital and oropharyngeal specimens. It is used in qualitative procedures for the isolation of Neisseria gonorrhoeae with suppression of most other gram-negative diplococci, gram-negative bacilli, gram-positive organisms, and yeast.