Membrane Filter Funnels, Microsart® 100/250 Funnels, Sartorius
Supplier: Sartorius
Certificates
About this item
PP housing.
- Reliable results-use a new, sterile funnel for each test for certain avoidances of cross contamination
- Time saving-kust change the funnel, rather than spending time sanitizing it
- Simpler handling-no more holding of hot funnels. And you can see when filtration has been completed, particularly useful when using manifolds in routine testing
- For filter diameter: 47 mm
Microsart® funnels are sterile 100 and 250 ml respectively plastic funnels. They allow quick performance of the filtrations required in the routine testing of water, food and beverages, pharmaceutical and cosmetic products.
Customers Who Bought This Also Bought