Orders LinkContactUsLinkComponent
Microsart® e.jet Pump
Microsart® e.jet Pump
Microsart® e.jet fluid pump, 4 Nl/min., 100/240 V (US plug)
Catalog # CA76517-796
Supplier:   Sartorius
Supplier #:  166MP4
Microsart® e.jet Pump
Catalog # CA76517-796
Supplier:  Sartorius
Microsart® e.jet Pump
Microsart® e.jet Pump
Microsart® e.jet fluid pump, 4 Nl/min., 100/240 V (US plug)
Price
$2,738.81
Log in for availability
Each
Product Details & Documents
The Microsart® e.jet is a laboratory vacuum pump which creates vacuum and concomitantly transfers the filtered liquid to waste.
  • For vacuum filtration in microbiological analysis
Microsart® e.jet is ideal for sample preparation in microbiological analysis achieving a trans membrane pressure of 600 mbar and a flow rate of 4.0 Nl/min (4.0 Normlitre water displacement by air in 1 minute). Constant flow rates and a defined maximum vacuum guarantees smooth and reliable filtration.
Specifications
  • Description:
    Microsart® e.jet fluid pump, 4 Nl/min.
  • Electrical:
    100/240 V (US plug)
Accessories
Product Information
  • Catalog #
    For Use With
    Availability
    Price
  • Quick connection set, 2 nipples (POM) on R³/₈" male thread and 2 couplings (PSU) on DN 10 hose nipple
    Quick connection set, 2 nipples (POM) on R³/₈" male thread and 2 couplings (PSU) on DN 10 hose nipple
    CA76517-940
    Microsart® e.jet pump
    Log in for availability
    Pack of 2 - $51.60