If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Size:36×50 µl
- Cat. no.:104013-340
DirectPlate™ JM109 chemically competent Escherichia coli cells are a perfect choice for researchers looking to simplify their transformation workflow by eliminating heat shock, lengthy incubations, and time-consuming outgrowth procedures. Simply mix and directly plate!
- No heat shock
- Save time (3 minutes transformation instead of a 90 minute transformation)
- High transformation efficiency
DirectPlate™ JM109 chemically competent Escherichia coli cells provide higher transformation efficiency than any competitor‘s similar product and are suitable for high-efficiency transformation in a wide variety of applications such as cloning and sub-cloning. Intact Genomics (ig®) DirectPlate™ competent cells offer simple, fast, and robust results for your DNA transformation needs. Genotype: endA1, recA1, gyrA96, thi, hsdR17 (rk–, mk+), relA1, supE44, Δ( lac-proAB), [F´ traD36, proAB, laqIqZΔM15].
The shipping and dry ice packaging cost is $45 for your entire order in the 48 contiguous United States.