CrossLab Sub-Microcells for UV-Vis and UV-Vis-NIR Systems, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Agilent offers a range of sub-micro rectangular cuvettes with optical path lengths of 5 or 10 mm, and a 20 mm height for compatibility with Cary UV systems.
- Sub-micro cells feature non transmitting, self masking black walls which improve sensitivity by eliminating stray light from measurements
- Fill volume reduced to approximately 20% of that used with a standard 10 mm path length rectangular cell
- Cells have 20 mm height for compatibility with Cary UV systems (excluding Cary 3500)
- Exceptional accuracy in terms of optical path length and parallelism
- Unique surface flatness of the optical windows
- High temperature stability
- High chemical resistance
- Outstanding measurement reproducibility
- Guaranteed transmission of >80% from 200 to 3500 nm (depending on the cell window material)
Sub-micro rectangular cuvettes (cells) reduce the fill volume to approximately 20% of that used with a standard 10 mm path length cell.
Frequently Bought Together