Orders LinkContactUsLinkComponent
Disintegration Baskets, Agilent Technologies
Disintegration Baskets, Agilent Technologies
Disintegration Baskets, Agilent Technologies
Certificates
Disintegration Baskets, Agilent Technologies
Disintegration Baskets, Agilent Technologies
10 Options Available Below
Product Details & Documents
Agilent offers disintegration baskets for the 100 automated disintegration apparatus.
Product Family Options
Product Information
  • Availability
    Price
  • CAAG37-4010
    3 Plastic Disks for 37-2030
    3 Plastic Disks for 37-2030
    Log in for availability
    Log in for availability
    Each
    Each - $309.00
    $309.00
    Each
       $309.00
    Supplier Number: 37-4010
  • CAAG37-3040
    Basket, 1-tube, 44 mm OD, 10-mesh, 1905 µm
    Basket, 1-tube, 44 mm OD, 10-mesh, 1905 µm
    Discontinued
    Supplier Number: 37-3040
  • CAAG37-2030
    Basket, 3-tube, 38 mm OD, 10-mesh, 1905 µm
    Basket, 3-tube, 38 mm OD, 10-mesh, 1905 µm
    Log in for availability
    Log in for availability
    Each
    Each - $924.00
    $924.00
    Each
       $924.00
    Supplier Number: 37-2030
  • CAAG37-3030
    Basket, 3-tube, 38 mm OD, 10-mesh, 1905 µm
    Basket, 3-tube, 38 mm OD, 10-mesh, 1905 µm
    Log in for availability
    Log in for availability
    Each
    Each - $915.00
    $915.00
    Each
       $915.00
    Supplier Number: 37-3030
  • CAAG37-3030V
    Basket, 3-tube, 38 mm OD, 10-mesh, 1905 µm, includes certificate
    Basket, 3-tube, 38 mm OD, 10-mesh, 1905 µm, includes certificate
    Log in for availability
    Log in for availability
    Each
    Each - $1,530.00
    $1,530.00
    Each
       $1,530.00
    Supplier Number: 37-3030V
  • CAAG37-3001
    Basket, 6-tube, 10-mesh, 1905 µm (USP)
    Basket, 6-tube, 10-mesh, 1905 µm (USP)
    Log in for availability
    Log in for availability
    Each
    Each - $812.00
    $812.00
    Each
       $812.00
    Supplier Number: 37-3001
  • CAAG37-2001
    Basket, 6-tube, 10-mesh, 1905 µm (USP)
    Basket, 6-tube, 10-mesh, 1905 µm (USP)
    Log in for availability
    Log in for availability
    Each
    Each - $818.00
    $818.00
    Each
       $818.00
    Supplier Number: 37-2001
  • CAAG37-2001V
    Basket, 6-tube, 10-mesh, 1905 µm (USP), includes certificate
    Basket, 6-tube, 10-mesh, 1905 µm (USP), includes certificate
    Log in for availability
    Log in for availability
    Each
    Each - $982.00
    $982.00
    Each
       $982.00
    Supplier Number: 37-2001V
  • CAAG37-3001V
    Basket, 6-tube, 10-mesh, 1905 µm (USP), includes certificate
    Basket, 6-tube, 10-mesh, 1905 µm (USP), includes certificate
    Log in for availability
    Log in for availability
    Each
    Each - $989.00
    $989.00
    Each
       $989.00
    Supplier Number: 37-3001V
  • CAAG37-2010
    Basket, 6-tube, 20-mesh, 864 µm
    Basket, 6-tube, 20-mesh, 864 µm
    Log in for availability
    Log in for availability
    Each
    Each - $873.00
    $873.00
    Each
       $873.00
    Supplier Number: 37-2010
  • CAAG37-3010
    Basket, 6-tube, 20-mesh, 864 µm
    Basket, 6-tube, 20-mesh, 864 µm
    Log in for availability
    Log in for availability
    Each
    Each - $973.00
    $973.00
    Each
       $973.00
    Supplier Number: 37-3010
  • CAAG37-3050
    Basket, 6-tube, 40-mesh, 381 µm, with screened cover
    Basket, 6-tube, 40-mesh, 381 µm, with screened cover
    Discontinued
    Supplier Number: -