Bond Elut Carbon & Carbon/NH₂, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Bond Elut Carbon is optimized to absorb pigments in food and small organic residues in water.
- Excellent retention for small organics, including those that are too polar to retain on C18 or polymeric SPE
- Removal of chlorophyll and other pigments leads to fewer chromatographic or mass interferences
- Broader retention and easier elution of analytes across the polarity range, for improved multiresidue analysis
- Carbon/NH2 and Carbon/PSA for enchanced polar compound removal
Bond Elut Carbon, Bond Elut Carbon/NH2, and Bond Elut Carbon/PSA cartridges are optimized for the absorption of pigments in food, fruits and vegetables, and small organic wastewater residues. Ultrapure graphitized carbon particles enable powerful retention mechanisms for a broad range of analytes. Careful manufacturing techniques result in lower carbon fines on the device wall.
Carbon/NH2 and Carbon/PSA contain NH2 and PSA, respectively, for enhanced removal of polar compounds.
Specifications
- Catalog No:
- CAAG126414
- CAAG12102201
- Polarity:
- Apolar stationary phase
- Apolar stationary phase