Bond Elut C18 OH, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Bond Elut C18 OH enhances retention of basic compounds and promotes metabolite fractionation.
- Silanol activity permits metabolite fractionation
- Tight QC tolerances deliver batch-to-batch reproducibility
- 150 Å pore size expands usage for higher molecular weight compounds
Bond Elut C18 OH is a nonendcapped version of the octadecyl bonded phases that enables the silanols on the silica surface to be more active. This low-load C18 phase has well-controlled silanol activity that permits the fractionation of metabolites and enhances retention of basic compounds compared to endcapped C18. The 150 Å pore size makes it a good choice for peptide extraction and higher molecular weight compounds.
Specifications
- Catalog No:
- CAAG12256040
- CAAG12102046
- Carbon load:
- 14.9%
- Endcapped:
- No
- Surface area:
- 300 m²/g
- Polarity:
- Apolar stationary phase
- Apolar stationary phase