CrossLab Solvent Filtration and Filter Membranes for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Improve LC system performance by removing contaminants and degassing the mobile phase.
- Degas eluents as particulates are removed
- Prevent the formation of spurious peaks within the detector due to solvent outgassing at the low-pressure end of the chromatograph
- Eliminate pump downtime caused by air locks and particulates in check valves
- Decrease piston wear
- Increase column life
- Increase solvent inlet lifetime
Mobile phase filters can improve the performance of your LC system by removing contaminants from solvents and degassing solvents in systems lacking on-line degassers.
Choose from a variety of different membrane filters compatible with normal- and reversed-phase mobile phases.
Related Information
Aglient Technologies Top Supplies to Optimize Your LC Workflow
Product Literature
Frequently Bought Together