ContactUsLinkComponent
VWR® Shell Vial and Plug Cap
VWR® Shell Vial and Plug Cap
Catalog # 66015-702A
Supplier:  VWR International
undefined
VWR® Shell Vial and Plug Cap
Catalog # 66015-702A
Supplier:  VWR International
Specifications
  • Cap included:
    Yes
  • Cap Material:
    PE
  • Capacity:
    1.00 ml
  • Color:
    Clear
  • Description:
    Shell vials with plug closure
  • Material:
    Borosilicate glass
  • Cat. no.:
    66015-702A
  • Diameter ×H:
    8×40 mm
  • Bottom Style:
    Flat bottom
Specifications
About this item
Designed for use in Alcott®, Waters®, Gilson®, or Shimadzu® autosamplers, the clear snap-plug closures of these shell vials can be pierced with standard autosampler syringes without vacuum formation.
  • Polyethylene plug with starburst center design eases syringe needle penetration
  • Convenient vial kits include equal quantities of vials and caps
  • Provides an economical option to autosampler vials
Manufactured from borosilicate glass. Vials and snap-plug closures packed separately in clear-pack, three chamber trays.