Slide-Sette Savers, Caplugs
Supplier: Caplugs
Jump to:
About this item
The Slide-Sette Saver is designed for shipping same patient pathology microscope slides and tissue/biopsy paraffin blocks in a single container.
- Each Slide-Sette Saver can be used to store specimens on a long-term basis
- Eliminate time spent looking for misplaced slides and/or paraffin blocks when the need for retrieval arises
- Available in multiple colors for various types of malignant tissue
The Slide-Sette Saver Storage Box is designed to hold multiple Slide-Sette Savers. It features a transparent polypropylene lid with a hinged bottom made of high-impact polystyrene.
9 Options Available Below
Product Details & Documents
The Slide-Sette Saver is designed for shipping same patient pathology microscope slides and tissue/biopsy paraffin blocks in a single container.
- Each Slide-Sette Saver can be used to store specimens on a long-term basis
- Eliminate time spent looking for misplaced slides and/or paraffin blocks when the need for retrieval arises
- Available in multiple colors for various types of malignant tissue
The Slide-Sette Saver Storage Box is designed to hold multiple Slide-Sette Savers. It features a transparent polypropylene lid with a hinged bottom made of high-impact polystyrene.
Product Family Options
Slide-Sette Savers
Product Information
- MaterialCapacityColorAvailabilityPrice
- Supplier Number: 240-3800-Z30
- Supplier Number: 240-3800-B30
- Supplier Number: 240-3800-K30
- Supplier Number: 240-3800-030
- Supplier Number: 240-3800-P30
- Supplier Number: 240-3800-U30
- Supplier Number: 240-3800-R30
- Supplier Number: 240-3800-Y30
Slide-Sette Saver Storage Box
Product Information
- MaterialCapacityColorAvailabilityPrice
- High-impact polystyrene base, polypropylene saver20 SlidesWhiteLog in for availabilityPack of 3$76.66Supplier Number: 240-3828-W80