About this item
A variety of tube cap options to meet needs. Test Tube Caps are overfitting caps for use with culture tubes.
- Two-position are only available in natural color
- Standard test tube stopper caps are available in a wide variety of sizes and colors
- Flat top stopper caps are designed to fit a variety of test tube sizes
Two-position caps are available non sterile or gamma irradiated for sterility.
Position one: Loose for aerobic culturing.
Position two: Tight for anaerobic culturing and leakproof sealing.