Screw Lid Specimen Containers, Caplugs
Supplier: Caplugs
Certificates
About this item
Screw lid specimen containers are designed for storing biological fluids and pathology specimens.
- Special thread design provides tight closure
- Graduations are calibrated in 10 ml increments
- Screw caps and most other surface areas provide ample space for identification and other markings
- All screw cap containers are also available gamma sterilized in individual packs with patient information labels
These multipurpose specimen containers are ideal for transporting samples, urine specimen collection and storing tissue specimens as well as for storage of pathological and cyto-histo specimens in 10% formalin.
Frequently Bought Together