Ward's® Lithographic Limestone
Lithographic limestone thin section
Catalog # 470331-478
Supplier: Avantor
Jump to:
Product Details & Documents
When minerals, rocks, and fossils cannot be accurately identified by macroscopic observation and testing, thin section slides can reveal the composition and structure of the specimens on a microscopic level.
Thin section slides are precisely ground to a thickness of 0.03 mm across the entire specimen area. Mineral and rock thin sections are mounted on 27×46 mm microscope slides, and protected by a glass coverslip. All thin section slides are labeled for reference.
Specifications
- Description:Lithographic limestone thin section