Orders LinkContactUsLinkComponent
SymLine® Continuous Level Control
SymLine® Continuous Level Control
Supplier:   SCAT AMERICAS, INC.
SymLine® Continuous Level Control
Certificates
SymLine® Continuous Level Control
SymLine® Continuous Level Control
2 Options Available Below
Product Details & Documents
Measuring filling levels continuously. Monitoring non-transparent canisters on a permanent basis is no problem with the continuous level control unit.
  • Automatic warning for 5 different filling levels freely adjustable, with accuracy of up to ±0,5 mm
  • Control box provides 5 freely configurable outlets to control external devices
  • Disc sensor for level control from non-conductive material is easy to install and adjustable in height
  • Rod sensor and canister with removable sleeve allow easy maintenance and cleaning
  • Canister made from electrostatic conductive HDPE material
The compact design and independent power supply allow simple handling and installation, also outside switch cabinets.
Product Family Options
Product Information
  • Availability
    Price
  • 76503-542
    Capacitive rod sensor for the level measuring with ATEX-license, II 2 G EEx ia IIC T1-T6, NAMUR
    Capacitive rod sensor for the level measuring with ATEX-license, II 2 G EEx ia IIC T1-T6, NAMUR
    Discontinued
    Supplier Number: 108178
  • 76503-654
    Floater, electronic, 139 mm
    Floater, electronic, 139 mm
    Log in for availability
    Log in for availability
    Each
    Each - $409.31
    $409.31
    Each
       $409.31
    Supplier Number: 160123
  • 76503-722
    Protective basket for floater, PP, suitable for all electro conductive canisters with floater
    Protective basket for floater, PP, suitable for all electro conductive canisters with floater
    Log in for availability
    Log in for availability
    Each
    Each - $111.85
    $111.85
    Each
       $111.85
    Supplier Number: 199013
  • 76504-062
    Set for continuous level control with Signalbox UM-O-Ex with 230 VAC
    Set for continuous level control with Signalbox UM-O-Ex with 230 VAC
    Discontinued
    Supplier Number: 502042
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Built-in Signalbox2; 2-channel (EU), for SafetyWasteCap and other electronic level controlers, touchless alarm-resetting fields for 2 containers, 2 switch inputs and 2 switch outputs for peripheral units incl. 230 V power supply cable with open end and clamp
    Built-in Signalbox2; 2-channel (EU), for SafetyWasteCap and other electronic level controlers, touchless alarm-resetting fields for 2 containers, 2 switch inputs and 2 switch outputs for peripheral units incl. 230 V power supply cable with open end and clamp
    76502-952
    Log in for availability
    Each - $1,607.97