SymLine® SwitchBox and 3-Way Ball Valve, SCAT
Automatically divert waste liquids into the reserve canister.
- Switchbox compatible to all electronic S.C.A.T. Europe and SymLine® level sensors
- Also compatible with the Signalbox and Built-in signalbox
- Many more applications, not only for waste disposal of HPLC systems
Improved safety through increased collecting volume, the reserve canister protects against overflowing, even for a longer time period. For example, waste can be disposed of over the weekend without operator supervision and without having to empty or change the full container.
Frequently Bought Together