Level Sensors, SCAT
These level sensors provide dependable protection from overflow when filling barrels. The float positions itself securely at the opening by its own weight while the barrel fills up. Available in different materials, therefore suitable for all chemicals.
- Integrated ventilation for safe pressure equalisation
- Fill level control for containers up to 200 liters
Float for easy level control when filling/refuelling drums. Robust and chemically resistant for heavy-duty use. Made of dissipative HD-PE material and compatible for all barrels with thread G ³/₄".
Frequently Bought Together