Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Color:Ultraviolet
- Description:Xite fluorescence flashlight system, 360 - 380 nm excitation, 415 nm longpass
- Cat. no.:76499-884
Specifications
About this item
Fluorescence Flashlight system that can be used in the lab or the field.
- Xite flashlights are not suitable for underwater use
- Two power levels, plus unique flashing mode and battery status indication
- Add-on diffuser for a broader, smoother beam
- Rechargeable high capacity lithium ion battery
The Xite Fluorescence Flashlight System includes single-wavelength fluorescence excitation flashlights with any of the five wavelengths in the NIGHTSEA line, paired with matching barrier filter glasses for maximum viewing contrast.
Dimensions: 13.5L×3.2Ø cm (5.3×1.26")