Anti-Evaporation oil, ibidi
Catalog # MSPP-50051
Supplier: IBIDI USA MS
CAS Number:
Specifications
- Description:Anti-Evaporation Oil, sterile, for cell culture
- Size:125 ml
- Cat. no.:MSPP-50051
- Supplier No.:50051
Specifications
About this item
A biocompatible silicone oil used to prevent medium evaporation in cell culture applications.
- Excellent reduction of medium evaporation
- Ready to use
- Dry-heat sterilized for direct use
- Viscosity optimized for cell culture
- Compatible with microscopy
- Compatible with all ibidi products and cell culture applications
Specifications
Appearance: Optically clear in dry atmosphere
Density (20°C): 1.05 g/ml
Viscosity (20°C): Ca. 200 mPa ∙ s
Refractive index nD: 1.453
Temperature stability: 121°C (dry-heat only)
Storage of product: Dark, room temperature