About this item
Sputocol is a self-contanted device for collecting, transporting and processing sputum and other infectious specimens.
- Ultra-clear polypropylene tubes
- Withstands 6000 RCF
- Inside edge of the funnel brings the specimen directly into the centrifuge tube and prevents contact with the tube rim
- Outside edge of the funnel is extended to provide a safe area for handling and removing
- The bottom lid detaches, functioning as a capping device
- Extra-wide base prevents tipping and a sturdy, protective carrier for the tube during transit
The large funnel provides a convenient 'target area'. Patients simply expectorate and reclose the funnel lid.