Reagent Reservoirs, Irish Life Sciences
Supplier: Irish Life Sciences
Certificates
About this item
These reagent reservoirs were designed especially for use with automated liquid handling platforms. They can also be used with manual systems.
- DNase/RNase free
- Polymer selection to overcome the surface tension of the liquid and minimize residual liquid
- Suitable for storage down to –80 °C
This reservoir has been developed precisely to conform to the footprint dimensions defined by SLAS/ANSI standards. This guarantees its compatibility with both manual and automated sample handling systems, readers, and microplate washers.
Certifications: Manufactured in ISO Class 8 Cleanroom.
Plate conforms to SLAS/ANSI standards.
Plate conforms to SLAS/ANSI standards.
Customers Who Bought This Also Bought