Deep Well Storage Plates, 96 Square Well, Irish Life Sciences
Supplier: Irish Life Sciences
Certificates
About this item
Square well plates made from medical grade polypropylene. Ideal for sample storage and collection.
- U and V bottom well format for optimal sample recovery
- Alphanumeric well coding
- High temperature and chemical resistance (–80 to +121 °C)
- Autoclavable
- RNase, DNase Free
- Stackable for use in plate and automation hotels
This plate has been developed precisely to conform to the footprint dimensions defined by SLAS/ANSI standards. This guarantees its compatibility with both manual and automated sample handling systems, readers, and microplate washers.
Certifications: Manufactured in ISO Class 8 Cleanroom.
Plate conforms to SLAS/ANSI standards.
Plate conforms to SLAS/ANSI standards.
Customers Who Bought This Also Bought