VWR® Wi-Fi Digital Data Logger (DDL)
76520-156
- Description:Wi-Fi digital data logger (DDL)
- Weight:1.1 kg (2.5 lbs.)
- Exterior W×D×H:203×127×76 mm (8×5×3")
- Cat. no.:76520-156
This Wi-Fi data logger is compliant with CDC guidelines. It includes a buffered probe in product simulated solution, °F/°C switchable readout, detachable dual probe (can be used with only one), audible and visual alarms, and temperature data via WIFI network. This device stores up to 3 years of data and includes a 3 year certificate of calibration.
- Material: Assorted materials