VWR® Wi-Fi Digital Data Logger (DDL) for Ultra Low Temperature Freezers (Dual Probe)
Catalog # 76520-148
Supplier: VWR International
Specifications
- Description:Wi-Fi digital data logger (DDL), for use with Ultra low temperature freezers (dual probe)
- Weight:1.1 kg (2.5 lbs.)
- Exterior W×D×H:203×127×76 mm (8×5×3")
- Cat. no.:76520-148
Specifications
About this item
This Wi-Fi data logger is compliant with CDC guidelines. It includes a buffered probe in product simulated solution, a detachable dual probe (can be used with only one), audible/visual alarms, and temperature data via Wi-Fi network. Device holds 3 years of data storage and comes with a 3 years certificate of calibration.
- Material: Assorted materials