ContactUsLinkComponent
CrossLab Inlet Liners for Agilent GC Instruments, Agilent Technologies
CrossLab Inlet Liners for Agilent GC Instruments, Agilent Technologies
  CA51905112025
CrossLab Inlet Liners for Agilent GC Instruments, Agilent Technologies
  CA51905112025
 :  5190-5112-025
  • Volume:
    870 µl
  • Description:
    Splitless, UI, Low-Frit Liner, 4 mm Int. Ø
  • Cat. no.:
    CA51905112025
Agilent provides a range of high-quality inlet liners for various third-party GC systems.
  • Designed, tested and manufactured by Agilent engineers to work perfectly with our instruments Superior performance
Agilent inlet liners provide the optimum environment for samples to be vaporized and mixed with the carrier gas in the inlet system. Agilent inlet liners provide the perfect mix of liner configurations and chemistries to solve a wide range of application challenges when using various GC systems. Choose from split, splitless, PTV, and other inlet liner designs, available in either the new, innovative Ultra Inert deactivation, or Agilent's popular original deactivation. With part and lot numbers silk-screened onto all Agilent liners, identification and re-ordering have never been easier.