Forceps for Membrane Filters, Sartorius
Catalog #: CA76517-784
Supplier: Sartorius
Specifications
- Description:Forceps for membrane filters, straight, with smooth, flat tips
- Cat. no.:CA76517-784
Specifications
About this item
These filter forceps are made of high grade stainless steel.
- Autoclavable, can be flamed
- With smooth, flat tips for a careful, firm hold of the membrane filter
Membrane filters should only be handled with suitable tweezers in order to avoid contamination which can result from hand contact.
Customers Who Bought This Also Bought