Orders LinkContactUsLinkComponent
Funnel, Sartorius
Funnel, Sartorius
Supplier:   Sartorius
Funnel, Sartorius
Supplier:  Sartorius
Certificates
Funnel, Sartorius
2 Options Available Below
Product Details & Documents
Stainless steel, autoclavable.
To use with filter holders for colony counting.
Ordering information: Supplied with funnel and clamp (lid and seal must be ordered separately).
Product Family Options
Product Information
  • Capacity
    Availability
    Price
  • CA76518-000
    Funnel with clamp
    Funnel with clamp
    500 ml
    Log in for availability
    Log in for availability
    Each
    Each - $564.13
    $564.13
    Each
       $564.13
    Supplier Number: 6981002
  • CA76518-008
    Funnel with clamp
    Funnel with clamp
    100 ml
    Log in for availability
    Log in for availability
    Each
    Each - $519.35
    $519.35
    Each
       $519.35
    Supplier Number: 6981065