ContactUsLinkComponent
Agilent Bio MAb, Agilent Technologies
undefined
Agilent Bio MAb, Agilent Technologies
  • Catalog No:
  • CAAG5190-2402
  • CAAG5190-2406
  • CAAG5190-2411
  • CAAG5190-2415
  • CAAG5190-2401
  • CAAG5190-2405
  • Max. pressure:
  • 689 bar
  • 413 bar
  • 413 bar
  • 275 bar
  • 689 bar
  • 413 bar
  • pH range:
  • 2-12
  • 2-12
  • 2-12
  • 2-12
  • 2-12
  • 2-12
  • Temperature limit:
  • 80 °C
  • 80 °C
  • 80 °C
  • 80 °C
  • 80 °C
  • 80 °C
  • Wetted materials:
  • Phosphate Buffer
  • Phosphate Buffer
  • Phosphate Buffer
  • Phosphate Buffer
  • Phosphate Buffer
  • Phosphate Buffer
Frequently Bought Together