Order Entry
Canada
Orders LinkContactUsLinkComponent
PL Multisolvent, Agilent Technologies

PL Multisolvent, Agilent Technologies

PL Multisolvent, Agilent Technologies
Certificates

2 Options Available Below

Product Details & Documents

PL Multisolvent GPC columns use polymer-coated silica to operate in a wide range of solvents.

  • The PL Multisolvent has the efficiency and robustness of silica with a special coating to eliminate ‘sticking’
  • Silica backbone provides the high strength necessary for high-efficiency particles and the rigid structure that eliminates pore swelling and collapse with solvent switching
  • High-resolution separation of polymers up to 100,000 Da, as well as additives, oligomers, and plasticizers

The PL Multisolvent family uses a revolutionary process to coat high-pore volume silica in an inert polymer. Columns separate low and moderate molecular weight polymers in solvents ranging from water to tetrahydrofuran (THF) without secondary interactions. The silica backbone provides excellent resolution and eliminates degradation caused by swelling during solvent transfer.

Specifications for Product Family

Specifications

  • Catalog No:
  • AGPL1515-3321
  • CAAGPL15153323
  • Max. pressure:
  • 150 bar
  • 2175.5 psi
  • pH range:
  • 1 - 14
  • 1 - 14
  • Temperature limit:
  • 90 °C
  • 90 °C
  • Wetted materials:
  • THF
  • THF

Specifications

Product Family Options

Product Information

  • Column Format
    Phase
    Particle Size
    Ø int.
    Length
    Packing material
    Availability
    Price
  • AGPL1515-3321
    PL Multisolvent 20
    PL Multisolvent 20
    Analytical
    PL Multisolvent 20
    2.7 µm
    4.60 mm
    150 mm
    Fully Porous
    Log in for availability
    Log in for availability
    Each
    Each - $2,380.00
    $2,380.00
    Each
       $2,380.00

    Supplier Number: PL1515-3321
    Specifications:
    More
    Specifications
    Cat. No.
    AGPL1515-3321
    Max. pressure
    150 bar
    pH range
    1 - 14
    Temperature limit
    90 °C
    Wetted materials
    THF
  • CAAGPL15153323
    PL Multisolvent 30
    PL Multisolvent 30
    Analytical
    PL Multisolvent 30
    2.7 µm
    4.60 mm
    150 mm
    Fully Porous
    Log in for availability
    Log in for availability
    Each
    Each - $2,240.00
    $2,240.00
    Each
       $2,240.00

    Supplier Number: PL1515-3323
    Specifications:
    More
    Specifications
    Cat. No.
    CAAGPL15153323
    Max. pressure
    2175.5 psi
    pH range
    1 - 14
    Temperature limit
    90 °C
    Wetted materials
    THF
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.