J&W Ultra Inert DB-FATWAX Columns, Agilent Technologies
A PEG-type GC column designed for analysis of FAMEs (e.g., fish oil) and fatty acids.
- Application-specific columns for FAMEs, FAEEs, and fatty acids
- Individually tested with a FAME mixture and Ultra Inert Wax test probe
- Reliable column-to-column inertness performance
- Improved peak shape for challenging polar compounds such as free fatty acids
- High polarity
- Equivalent to USP G16
- Bonded and crosslinked phase is solvent rinseable and tolerates aqueous injections
Agilent J&W DB-FATWAX Ultra Inert (UI) is an application-specific polyethylene glycol (PEG)-type GC column designed for the separation of fatty acid methyl esters (FAMEs), fatty acid ethyl esters (FAEEs), and fatty acids. This column is tested with a FAME mixture to ensure reproducible FAME equivalent chain length (ECL) values, proper identification of important FAMEs such as EPA, DPA, and DHA, and resolution of key pairs of FAMEs.
- Catalog No:
- CAAGG390363009
- CAG3903-63008
- CAAGG390363007
- Polarity:
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
Frequently Bought Together