J&W DB-FastFAME Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Speeds up FAME analysis without sacrificing resolution for critical cis and trans FAME isomers.
- Cyanopropyl phase (G48) engineered for the fast and selective separation of FAMEs
- Individually tested with a FAME mixture to ensure reproducible FAME equivalent chain length (ECL) values
- Meets the requirements of AOAC, AOCS, and IOC regulatory methods
- Bonded, crosslinked, and solvent rinseable
- Also available in Intuvo configuration
Agilent J&W DB-FastFAME is a cyanopropyl phase engineered for the fast and selective separation of polysaturated and unsaturated FAMEs-even challenging cis and trans FAME isomers—without sacrificing the resolution generally achieved with traditional 100 meter long columns. With DB-FastFAME, it is possible to separate traditional FAMEs in under 8 minutes, including C18:1 and C18:2 cis-trans isomers and FAMEs commonly found in milk fat, vegetable oil, and fish oil, such as DPA and EPA.
Specifications
- Catalog No:
- CAAGG390363012
- CAAGG390363013
- CAAGG390363011
- Polarity:
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase