CrossLab Gas Tight Syringes, Agilent Technologies
Catalog # CASG003987
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- Volume:25 µl
- Description:Syringe, SGE, gas-tight
- Cat. no.:CASG003987
- Needle Length:50 mm
- Needle Type:Fixed
- Needle Gauge:23
- Tip Type:Cone
Specifications
About this item
Gas tight syringes for dispensing gases and liquids.
- Precisely made for optimal and repeatable performance through accurate volume delivery
These gas tight syringes are constructed of borosilicate glass and PTFE plunger tips. An interference fit between the plunger tip and the barrel enhances the gas sealing properties of the syringe.