CrossLab Tubing for Cary UV Accessories, Agilent Technologies
Catalog # CA9910052900
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- Ø int.:4.76 mm
- Description:Tubing, peristaltic pump kit for the Cary Routine Sampler Accessory
- Length:118 mm
- Material:Silicone
- Cat. no.:CA9910052900
Specifications
About this item
Tubing spares and kits for Cary UV-Vis spectrophotometer instruments and accessories.
- Designed, tested, and manufactured with the same attention to detail you expect from Agilent instruments
- Using non-Agilent supplies can considerably shorten lifetime and increases potential baseline noise
These allow you to replace peristaltic pump tubing and inlet/outlet connections for flow cells to help you keep your system operating at peak performance.