ContactUsLinkComponent
NIGHTSEA Stereo Microscope Fluorescence Adapter for Keyence VHX Series, Electron Microscopy Sciences
NIGHTSEA Stereo Microscope Fluorescence Adapter for Keyence VHX Series, Electron Microscopy Sciences
Catalog # 76499-858
undefined
NIGHTSEA Stereo Microscope Fluorescence Adapter for Keyence VHX Series, Electron Microscopy Sciences
Catalog # 76499-858
Supplier Number:  SFA-KFI-IND
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Keyence FI system
  • Cat. no.:
    76499-858
Specifications
About this item
NIGHTSEA Model SFA Fluorescence Adapter system adds versatile fluorescence imaging to the VHX series.
Keyence offers a variety of lenses for imaging at different scales. NIGHTSEA offers several variants of the fluorescence adapter system that work with the most commonly used Keyence lenses – the ZST, Z100, Z50, Z20, and Z00.
NIGHTSEA implements these for the Keyence system with high intensity LED light sources (available in five excitation wavelength ranges), and emission barrier filters that can be added to the Keyence lenses easily and non-invasively.