EDVOTEK® Personal PCR LabStation™
Catalog # 470336-112
Supplier: Edvotek
Specifications
- Gradient:-
- Description:Personal PCR LabStation™
- Cat. no.:470336-112
Specifications
About this item
Personal PCR for the classroom! Ideal for individual research and small group projects.
- Supports 16 students
Personal PCR for the classroom! LabStation™ includes a 16-well personal PCR machine, two electrophoresis gel boxes, power supply, two variable pipettes, blue light transilluminator, and water bath!
Delivery: Includes 1× model 540 EdvoCycler™ Jr. (16×0.2 ml); 2× model 502-504 M12 Complete™ Package (14×7 cm Tray and 7×7 cm Trays (2)); 1 x model 509 DuoSource™ Power Supply (100 or 150 V, for 1 to 2 units); 2× model 590 Variable MicroPipette (5 to 50 µl); 1× model 557 TruBlu™ LED Transilluminator; 1× model 539 3 L Water Bath.