HardyCHROM™ Candida + Auris, Hardy Diagnostics
Catalog # CA76509-142
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Specifications
- Description:Candida + auris plates
- Size:10/pack
- Cat. no.:CA76509-142
- Plate Size:15×100 mm
Specifications
About this item
Hardy Diagnostics HardyCHROM™ Candida+auris is recommended for the selective isolation and differential identification of Candida species.
- Candida auris produces white colonies with teal to teal-green 'bullseye' centers that are positive for UV fluorescence for easy differentiation from other Candida species
- Colonies stand out with bright, vivid color development against a white background for effortless interpretation
- Useful as an aid for differentiating other clinically relevant Candida species (C. albicans, C. tropicalis, C. krusei)
- Inhibitory to most bacteria common to mixed specimens (e.g. skin, throat, axilla, urogenital tract, or rectal swabs, and more)
- Most strains will exhibit colors within 48 hours
The medium allows for the differentiation of C. albicans, C. tropicalis, C. krusei and C. auris based on colony morphology, color, and positive UV fluorescence.
The HardyCHROM™ plates use optically clear, heavy-duty polystyrene for distortion-free viewing and reduced breakage during shipping. Receive your pack in a 'breathable' sleeve of 10 plates per bag that prevents the buildup of condensation and excess moisture.