Stacking Banana Jack Snaps, 10 mm Socket, Transforming Technologies
Supplier: Transforming Technologies
Certificates
About this item
Universal stacking snap adapters add a banana jack to a 10 mm snap or stud, typically on an ESD mat.
- Universal snap adapter can connect to both studs and sockets
- Adds a banana jack to a 10 mm snap or stud, typically on an ESD mat
- Easily connect a wrist strap, ground cord, or constant monitor
- Great for grounding mats on carts
The banana jacks are available with both a 10 mm male stud and a 10 mm female socket. The banana jack is compatible and interchangeable with standard banana jacks.
Frequently Bought Together