Socket Connection, 10 mm to 10 mm, Transforming Technologies
Catalog # 76489-638
Supplier: Transforming Technologies
Specifications
- Description:10 mm socket to socket connection
- Cat. no.:76489-638
Specifications
About this item
This Snap Adapter Set is compatible and interchangeable with all major snap manufactures.
- Steel adapter has a double 10 mm stud construction
- Used to convert 10 mm snaps, primarily with anti-static matting and ground cords
- Snap adapter connects a 10 mm socket to a 10 mm socket
Also available in a stud-to-stud connection with the CS0128-stud.