ContactUsLinkComponent
Interconnect Cord, with 2 Banana Plugs, Transforming Technologies
Interconnect Cord, with 2 Banana Plugs, Transforming Technologies
Catalog # 76489-586
undefined
Interconnect Cord, with 2 Banana Plugs, Transforming Technologies
Catalog # 76489-586
Supplier Number:  CC3000NR
Specifications
  • Description:
    Interconnect cord, with 2 banana plugs
  • Length:
    8'
  • Cat. no.:
    76489-586
Specifications
About this item
Two standard banana plugs allow the cord to connect two devices such as mats or ground cords that feature banana jacks.
  • Use to connect two items together
  • Each end terminates with a standard banana plug
  • Length is 8'
  • No resistor
The Interconnect Cord features 2.5 mm polyurethane insulation that offers excellent cord life.