ContactUsLinkComponent
Conductive Trash Can Dolly, Transforming Technologies
Conductive Trash Can Dolly, Transforming Technologies
Catalog # 76490-264
Conductive Trash Can Dolly, Transforming Technologies
Catalog # 76490-264
Supplier Number:  WBASDM
Specifications
  • Description:
    Conductive trash dolly
  • Cat. no.:
    76490-264
Specifications
About this item
The cart provides a user-friendly solution for heavy ESD trash containers.
  • Industrial aluminum construction
  • Wider than plastic dolly
  • Solid aluminum hubs
The dolly locks on to the 44 gallon WBAS180 Transforming Technologies container. The 5 heavy-duty casters provide stability for loads up to 350 lbs., and two casters are conductive to provide a path to ground for ESD charges.