PEGuard Impervious Shoe Covers, Apex Aseptic Products
Supplier: APEX ASEPTIC PRODUCTS, LLC.
Certificates
About this item
PEGuard Impervious Shoe Covers are made of durable, skid-resistant PE coated non-woven fabrics. These garments are impervious and lint-free.
- Durable, tear-resistant, water-resistant
- Anti-skid treated to help prevent accidental falls
- Latex-free, Silicone-free, and nonsterile
- Low particle shedding, category 1 per Helmke Drum Test
- Manufactured in controlled environment
- Reasonable cost, affordable for everyday use
PEGuard Impervious Shoe Covers are made with specially treated, heavy-duty PE coated non-woven fabrics. They are ideal for use in pharmaceutical manufacturing facilities, animal labs, research centers, medical establishments, and cleanroom environments.
Certifications: Manufacturing facility is ISO 9001 and ISO 13485 certified. FDA registered and inspected.
Packaging: Shoe covers are packaged 50 per bag, 300 per case.
Customers Who Bought This Also Bought