Orders LinkContactUsLinkComponent
Disposable Forceps
Disposable Forceps
Supplier:  BURKLE INC
Certificates
Disposable Forceps
Disposable Forceps
9 Options Available Below
Product Details & Documents
These disposable forceps are suitable for precise lab work.
  • PS, white or blue
  • Individually packaged
    Both sterile and non sterile options available
  • For single use
  • Clean room manufactured (clean room class 7)
  • Overall length 130 mm (5.12")
They are ideal for grasping, lifting and moving small objects, for example fine tissue, small items or solid samples.
 : Complies with EU food requirements and FDA regulations.
Product Family Options
Product Information
  • Color
    Overall length
    Sterile
    Availability
    Price
  • 76501-438
    Disposable forceps, broad tip
    Disposable forceps, broad tip
    Blue
    130 mm (5.21")
    +
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $173.43
    $173.43
    Pack of 100
       $173.43
    Supplier Number: 5378-1052
  • 76501-430
    Disposable forceps, broad tip
    Disposable forceps, broad tip
    White
    130 mm (5.21")
    +
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $171.90
    $171.90
    Pack of 100
       $171.90
    Supplier Number: 5378-1042
  • 76501-432
    Disposable forceps, broad tip
    Disposable forceps, broad tip
    White
    130 mm (5.21")
    -
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $106.20
    $106.20
    Pack of 100
       $106.20
    Supplier Number: 5378-0042
  • 76501-436
    Disposable forceps, pointed
    Disposable forceps, pointed
    Blue
    130 mm (5.21")
    +
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $176.55
    $176.55
    Pack of 100
       $176.55
    Supplier Number: 5378-1051
  • 76501-426
    Disposable forceps, pointed
    Disposable forceps, pointed
    White
    130 mm (5.21")
    +
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $176.55
    $176.55
    Pack of 100
       $176.55
    Supplier Number: 5378-1041
  • 76501-428
    Disposable forceps, pointed
    Disposable forceps, pointed
    White
    130 mm (5.21")
    -
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $106.20
    $106.20
    Pack of 100
       $106.20
    Supplier Number: 5378-0041
  • 76501-434
    Disposable forceps, pointed/angled
    Disposable forceps, pointed/angled
    Blue
    130 mm (5.21")
    +
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $176.55
    $176.55
    Pack of 100
       $176.55
    Supplier Number: 5378-1050
  • 76501-422
    Disposable forceps, pointed/angled
    Disposable forceps, pointed/angled
    White
    130 mm (5.21")
    +
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $173.43
    $173.43
    Pack of 100
       $173.43
    Supplier Number: 5378-1040
  • 76501-424
    Disposable forceps, pointed/angled
    Disposable forceps, pointed/angled
    White
    130 mm (5.21")
    -
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $106.20
    $106.20
    Pack of 100
       $106.20
    Supplier Number: 5378-0040