ContactUsLinkComponent
LaMotte® Phosphate, Low Range
LaMotte® Phosphate, Low Range
Catalog # 470335-414
Supplier:  LaMotte
CAS Number:  
undefined
LaMotte® Phosphate, Low Range
Catalog # 470335-414
Supplier:  LaMotte
Supplier Number:  3121-CN-02
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Water test kit, Phosphate, low range
  • Environmentally Preferable:
  • Cat. no.:
    470335-414
Specifications
About this item
Pre-calibrated test tubes and labware give fast, accurate results in the classroom or in the lab.
  • Range: 0 to 2 ppm
  • Material for 50 tests
  • Compact, waterproof carrying case contains kit
  • Instructions included
  • Pre-calibrated test tubes