Order Entry
Canada
ContactUsLinkComponent
HyClone™ FetalClone™ I serum, U.S. origin, HyClone products (Cytiva)
HyClone™ FetalClone™ I serum, U.S. origin, HyClone products (Cytiva)
Catalog # CA76324-880
Supplier:  Cytiva
undefined
HyClone™ FetalClone™ I serum, U.S. origin, HyClone products (Cytiva)
Catalog # CA76324-880
Supplier:  Cytiva
Supplier Number:  3008003IR25-40

Specifications

  • Serum format:
    Frozen
  • Description:
    HyClone FetalClone I serum, US Origin
  • Size:
    500 ml
  • Origin:
    US
  • Hemoglobin:
    ≤25 mg/dl
  • Endotoxin:
    ≤10EU/ml
  • Storage temperature:
    Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
  • Cat. no.:
    CA76324-880

Specifications

About this item

HyClone™ FetalClone® I is a foetal bovine serum alternative optimised for the growth of hybridomas with comparable IgG levels to those found in FBS.

  • IgG levels comparable with those found in FBS
  • Demonstrated cell growth performance equivalency to FBS
  • High lot-to-lot consistency
  • Low endotoxin and hemoglobin levels

Sterile filtered using three sequential 100 nm (0.1 µm) pore-size rated filters. Each lot undergoes extensive quality performance and analytical testing.