Borosil® Reusable Class A Mohr (Measuring) Pipettes (Pipettes) with Individual Serial Numbers, Foxx Life Sciences®
Supplier: Foxx Life Sciences
Certificates
About this item
These Borosil brand pipettes are made of specially-selected heavy wall machine-drawn borosilicate accurate bore glass tubing.
- Markings are made in permanent amber stain diffused into the surface of the glass for better visibility and durability
- Graduated for delivery from zero mark to the last graduation mark
They have precision formed jets, and bevel ground ends that resist chipping. The graduation marks are made in durable white enamel or amber stain, resistant to normal laboratory reagents and repeated washing. All the Pipettes are individually measured, and then marked. The uniformity of bore ensures accuracy between any two readings. The Pipettes can be autoclaved and, if necessary, can be flamed for sterilization.
Certifications: Complies with IS, ISO / DIN 835.
Delivery: Supplied with individual or bulk calibration certificate showing actual volume dispensed and providing traceability to national standards.
Frequently Bought Together