Cadaver Cover for JC100 Body Tray and Frame, Mortech®
76498-638
: Mopec
- Color:White
- Description:Cadaver cover for JC100 body tray and frame
- Cat. no.:76498-638
This cadaver cover is designed specifically for use with the JC100 body tray and frame.
- For use with JC100 body tray and frame
- Color: white
Cadaver covers allow for the discrete transport of a patient from the hospital room to the mortuary. After placing the body and the frame on the carrier, place cover over the frame. The rectangular shape of the cover conceals the body, giving the illusion of a standard gurney while hiding the cadaver.